Gene Transcripts

Gene Symbol
HYAL4
Entrez Gene ID
23553
Gene Name
hyaluronoglucosaminidase 4
Gene Aliases
CSHY
Chromosome Location
Chr. 7: 123763708 - 123877481 on Build GRCh38
UniGene ID
Hs.28673
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012269.2 NP_036401.2 3 1170
XM_011515990.2 XP_011514292.1 3 1246
XM_011515991.2 XP_011514293.1 3 828
XM_017011911.1 XP_016867400.1 4 873
GenBank mRNA AB470346.1 - 2 602
AF009010.1 - 3 1173
BC104788.1 - 2 608 8143791
BC104790.1 - 2 608 8143793

View Full Product Details