Gene Transcripts

Gene Symbol
TMEM50A
Entrez Gene ID
23585
Gene Name
transmembrane protein 50A
Gene Aliases
IFNRC, SMP1
Chromosome Location
Chr. 1: 25338298 - 25362361 on Build GRCh38
UniGene ID
Hs.705699
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014313.3 NP_055128.1 3, 4 369
XM_005245817.1 XP_005245874.1 3, 4 333
XM_011541159.1 XP_011539461.1 3, 4 563
GenBank mRNA AF081282.1 - 3, 4 296
AF087880.1 - 3, 4 357
AK299180.1 - 3, 4 333
AK312588.1 - 3, 4 333
AL136627.1 - 3, 4 351
AY071927.1 - 3, 4 346
AY358650.1 - 3, 4 332
BC007341.2 - 3, 4 317 3688672
BC066908.1 - 3, 4 308 4817784
JX644905.1 - 3, 4 369
JX644906.1 - 3, 4 369

View Full Product Details