Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001242854.1 | NP_001229783.1 | 5 | 813 | |
NM_001242855.1 | NP_001229784.1 | 4 | 573 | ||
NM_001242856.1 | NP_001229785.1 | 3 | 454 | ||
NM_012402.3 | NP_036534.1 | 5 | 674 | ||
XM_005252840.4 | XP_005252897.1 | 5 | 578 | ||
GenBank mRNA | AK026390.1 | - | 5 | 506 | |
AK300855.1 | - | 5 | 674 | ||
AK301856.1 | - | 4 | 528 | ||
AK301972.1 | - | 5 | 615 | ||
AK304512.1 | - | 3 | 321 | ||
AK311086.1 | - | 4 | 1156 | ||
BC000392.2 | - | 5 | 496 | 2821666 | |
BM928413.1 | - | 641 | |||
U52522.1 | - | 5 | 490 | ||
X97567.1 | - | 3 | 309 |