Gene Transcripts

Gene Symbol
QPCT
Entrez Gene ID
25797
Gene Name
glutaminyl-peptide cyclotransferase
Gene Aliases
GCT, QC, sQC
Chromosome Location
Chr. 2: 37344610 - 37373322 on Build GRCh38
UniGene ID
Hs.79033
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012413.3 NP_036545.1 3 503
GenBank mRNA AK290605.1 - 3 485
BC020456.1 - 3 404
BC036721.1 - 3 382 5224222
BC047756.2 - 3 480 6058335
BI755122.1 - 3 538
X67731.1 - 2 185
X71125.1 - 3 392

View Full Product Details