Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015631.5 | NP_056446.4 | 5 | 916 | |
XM_005269690.1 | XP_005269747.1 | 5 | 758 | ||
XM_011539627.1 | XP_011537929.1 | 5 | 758 | ||
XM_011539628.1 | XP_011537930.1 | 5 | 758 | ||
GenBank mRNA | AK075357.1 | - | 5 | 701 | |
AK098295.1 | - | 4 | 894 | ||
AL050022.1 | - | 5 | 700 | ||
AY359022.1 | - | 5 | 680 | ||
BC009494.2 | - | 2 | 76 | ||
BC061582.1 | - | 5 | 587 | ||
BC068449.1 | - | 5 | 916 | 5298627 | |
DQ278872.1 | - | 5 | 916 |