Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012158.2 | NP_036290.1 | 2 | 627 | |
XM_005266336.1 | XP_005266393.1 | 2 | 410 | ||
XM_017020538.1 | XP_016876027.1 | 2 | 611 | ||
GenBank mRNA | AF126028.1 | - | 1 | 231 | |
AF129532.1 | - | 2 | 294 | ||
AK314442.1 | - | 2 | 595 | ||
AL833187.1 | - | 2 | 575 | ||
BC072448.1 | - | 2 | 581 | 6166244 |