Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017417.1 | NP_059113.1 | 9 | 1660 | |
GenBank mRNA | AJ271385.1 | - | 9 | 1660 | |
AJ505959.1 | - | 2 | 188 | ||
BC140888.1 | - | 9 | 1626 | 9021756 | |
BC140889.1 | - | 9 | 1626 | 9021757 |