Gene Transcripts
- Gene Symbol
- OR7C2
- Entrez Gene ID
- 26658
- Gene Name
- olfactory receptor family 7 subfamily C member 2
- Gene Aliases
- CIT-HSP-87M17, OR19-18, OR7C3
- Chromosome Location
- Chr. 19: 14941489 - 14942448 on Build GRCh38
- UniGene ID
- Hs.589620
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012377.1 |
NP_036509.1
|
1 |
778 |
|
View Full Product Details