Gene Transcripts
- Gene Symbol
- OR2T1
- Entrez Gene ID
- 26696
- Gene Name
- olfactory receptor family 2 subfamily T member 1
- Gene Aliases
- OR1-25
- Chromosome Location
- Chr. 1: 248405995 - 248407104 on Build GRCh38
- UniGene ID
- Hs.626885
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030904.1 |
NP_112166.1
|
1 |
105 |
|
View Full Product Details