Gene Transcripts

Gene Symbol
DNAI1
Entrez Gene ID
27019
Gene Name
dynein axonemal intermediate chain 1
Gene Aliases
CILD1, DIC1, ICS1, PCD
Chromosome Location
Chr. 9: 34458752 - 34520989 on Build GRCh38
UniGene ID
Hs.112667
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281428.1 NP_001268357.1 16 1771
NM_012144.3 NP_036276.1 16 1759
XM_006716758.3 XP_006716821.1 15 1402
XM_011517846.1 XP_011516148.1 16 1771
XM_011517847.2 XP_011516149.1 16 1771
XM_011517849.1 XP_011516151.1 16 1771
GenBank mRNA AF091619.1 - 16 1676
AK290696.1 - 16 1683
AK302186.1 - 13 1302
AK314639.1 - 16 1671
BC030583.2 - 16 1687 4837991

View Full Product Details