Gene Transcripts
- Gene Symbol
- SNX11
- Entrez Gene ID
- 29916
- Gene Name
- sorting nexin 11
- Gene Aliases
- Chromosome Location
- Chr. 17: 48107533 - 48123064 on Build GRCh38
- UniGene ID
- Hs.15827
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013323.2 |
NP_037455.2
|
2, 3 |
316 |
|
NM_152244.1 |
NP_689450.1
|
3, 4 |
403 |
|
XM_005257260.3 |
XP_005257317.1
|
2, 3 |
83 |
|
XM_011524697.2 |
XP_011522999.1
|
3, 4 |
186 |
|
GenBank mRNA |
AF121861.1 |
- |
2, 3 |
336 |
|
AK023932.1 |
- |
3, 4 |
403 |
|
AK091852.1 |
- |
3, 4 |
301 |
|
AK296569.1 |
- |
3, 4 |
385 |
|
BC000768.2 |
- |
2, 3 |
133 |
2963905 |
BC103721.1 |
- |
2, 3 |
122 |
6162638 |
BT006723.1 |
- |
1, 2 |
49 |
|
View Full Product Details