Gene Transcripts

Gene Symbol
HUNK
Entrez Gene ID
30811
Gene Name
hormonally up-regulated Neu-associated kinase
Gene Aliases
Chromosome Location
Chr. 21: 31873315 - 32004064 on Build GRCh38
UniGene ID
Hs.109437
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014586.1 NP_055401.1 6 1279
XM_011529537.2 XP_011527839.1 6 943
GenBank mRNA AJ271722.1 - 6 1279

View Full Product Details