Gene Transcripts

Gene Symbol
APH1A
Entrez Gene ID
51107
Gene Name
aph-1 homolog A, gamma-secretase subunit
Gene Aliases
6530402N02Rik, APH-1, APH-1A, CGI-78
Chromosome Location
Chr. 1: 150265399 - 150282291 on Build GRCh38
UniGene ID
Hs.108408
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077628.2 NP_001071096.1 2 649
NM_016022.3 NP_057106.2 2 649
NR_045035.1 2 649
GenBank mRNA AF151835.1 - 2 351
AF508787.1 - 2 307
AK027879.1 - 2 419
AK075295.1 - 2 368
AK075452.1 - 2 457
AK298876.1 - 2 428
AK314490.1 - 2 419
AX775955.1 - 2 385
AX775957.1 - 2 439
AY113698.1 - 2 307
AY113699.1 - 2 307
AY358951.1 - 2 453
BC001230.2 - 2 419 3457593
BC008732.2 - 2 406 3162330
BC009501.2 - 2 440 4040230
BC015568.2 - 2 408 3356401
BC017699.1 - 2 385 4177580
BC020590.1 - 2 439 4420942

View Full Product Details