Gene Transcripts

Gene Symbol
SAR1B
Entrez Gene ID
51128
Gene Name
secretion associated Ras related GTPase 1B
Gene Aliases
ANDD, CMRD, GTBPB, SARA2
Chromosome Location
Chr. 5: 134601149 - 134632843 on Build GRCh38
UniGene ID
Hs.432984
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033503.2 NP_001028675.1 8 835
NM_016103.3 NP_057187.1 7 716
XM_017009521.1 XP_016865010.1 7 723
GenBank mRNA AF087850.1 - 7 753
AF092130.1 - 7 732
AK056821.1 - 4 2550
AK223452.1 - 7 701
AL512710.1 - 7 1751
BC002847.2 - 8 766 3636540
BC093034.1 - 7 716 30528518

View Full Product Details