Gene Transcripts

Gene Symbol
SLC15A3
Entrez Gene ID
51296
Gene Name
solute carrier family 15 member 3
Gene Aliases
OCTP, PHT2, PTR3
Chromosome Location
Chr. 11: 60937083 - 60952081 on Build GRCh38
UniGene ID
Hs.237856
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016582.2 NP_057666.1 7, 8 1836
NR_027391.1 6, 7 1667
XM_011545095.2 XP_011543397.1 8, 9 2182
GenBank mRNA AB020598.1 - 7, 8 1836
AF135600.1 - 7, 8 1729
AK127216.1 - 2, 3 4291
BC025710.1 - 6, 7 1527 5224878
BC037974.1 - 7, 8 1696 5213632
BU731752.1 - 1 295

View Full Product Details