Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001242923.1 | NP_001229852.1 | 3 | 175 | |
NM_016091.3 | NP_057175.1 | 3 | 175 | ||
XM_005261625.1 | XP_005261682.1 | 3 | 130 | ||
XM_006724260.3 | XP_006724323.1 | 3 | 773 | ||
XM_017028814.1 | XP_016884303.1 | 3 | 130 | ||
GenBank mRNA | AF077207.1 | - | 3 | 111 | |
AF083243.1 | - | 3 | 130 | ||
AF109359.1 | - | 3 | 112 | ||
AK056129.1 | - | 3 | 679 | ||
AK093540.1 | - | 2 | 599 | ||
AK129648.1 | - | 3 | 128 | ||
AK222496.1 | - | 3 | 130 | ||
AK222529.1 | - | 3 | 128 | ||
AK293629.1 | - | 3 | 130 | ||
AK298776.1 | - | 3 | 106 | ||
AK301323.1 | - | 3 | 129 | ||
AK302346.1 | - | 3 | 130 | ||
AK315533.1 | - | 3 | 136 | ||
AL357455.1 | - | 3 | 103 | ||
BC001101.2 | - | 3 | 113 | 3507964 | |
BC007510.2 | - | 3 | 113 | 2959747 | |
BC029265.1 | - | 3 | 115 | 5110941 | |
CR456436.1 | - | 3 | 236 | ||
DA051005.1 | - | 3 | 175 | ||
DA663575.1 | - | 3 | 136 |