Gene Transcripts

Gene Symbol
SNX9
Entrez Gene ID
51429
Gene Name
sorting nexin 9
Gene Aliases
SDP1, SH3PX1, SH3PXD3A, WISP
Chromosome Location
Chr. 6: 157716046 - 157945077 on Build GRCh38
UniGene ID
Hs.191213
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016224.4 NP_057308.1 5 611
XM_005267015.2 XP_005267072.1 7 689
XM_011535886.2 XP_011534188.1 2 150
GenBank mRNA AF001629.1 - 5 416
AF121859.1 - 5 371
AF131214.1 - 5 390
AF172847.1 - 5 491
AK026637.1 - 2 156
AK127345.1 - 2 466
AK314360.1 - 5 445
BC005022.1 - 5 518 3832234
BG766499.1 - 5 521
DA336311.1 - 5 557

View Full Product Details