Gene Transcripts

Gene Symbol
HIKESHI
Entrez Gene ID
51501
Gene Name
Hikeshi, heat shock protein nuclear import factor
Gene Aliases
C11orf73, HLD13, HSPC138, HSPC179, L7RN6, OPI10
Chromosome Location
Chr. 11: 86302211 - 86345943 on Build GRCh38
UniGene ID
Hs.283322
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322404.1 NP_001309333.1 2 275
NM_001322407.1 NP_001309336.1 3 449
NM_001322409.1 NP_001309338.1 3 379
NM_016401.3 NP_057485.2 2 291
NR_136324.1 2 275
XM_017017914.1 XP_016873403.1 2 281
XM_017017915.1 XP_016873404.1 2 68
GenBank mRNA AF161487.1 - 2 275
AF161527.1 - 2 274
AK001447.1 - 2 262
AK223198.1 - 2 262
AK298763.1 - 2 251
BC001677.2 - 2 164 2820772
BC006476.2 - 2 164 2820772
BC015991.2 - 2 256 2820691
BC018080.1 - 2 225 4798958
BC021621.1 - 2 256 2820691
BX332668.2 - 2 128

View Full Product Details