Gene Transcripts

Gene Symbol
CCSER2
Entrez Gene ID
54462
Gene Name
coiled-coil serine rich protein 2
Gene Aliases
FAM190B, Gcap14, KIAA1128, bA486O22.1
Chromosome Location
Chr. 10: 84328558 - 84518521 on Build GRCh38
UniGene ID
Hs.461988
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284240.1 NP_001271169.1 2 1435
NM_001284241.1 NP_001271170.1 2 1435
NM_018999.3 NP_061872.2 2 1435
XM_005269905.3 XP_005269962.1 2 1466
XM_005269906.3 XP_005269963.1 2 1466
XM_011539870.2 XP_011538172.1 2 1466
XM_011539871.2 XP_011538173.1 2 1466
XM_011539872.2 XP_011538174.1 2 1451
XM_017016340.1 XP_016871829.1 2 1355
XM_017016341.1 XP_016871830.1 2 1451
XM_017016342.1 XP_016871831.1 2 1433
XM_017016343.1 XP_016871832.1 1 1196
GenBank mRNA AB032954.1 - 1 438
AF241785.1 - 1 924
AK024324.1 - 1 1159
AK298966.1 - 2 1361
BC030528.1 - 2 1333 5208702
CR991891.1 - 1 88

View Full Product Details