Gene Transcripts

Gene Symbol
CPVL
Entrez Gene ID
54504
Gene Name
carboxypeptidase, vitellogenic like
Gene Aliases
HVLP
Chromosome Location
Chr. 7: 28995630 - 29146537 on Build GRCh38
UniGene ID
Hs.233389
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019029.2 NP_061902.2 8 879
NM_031311.3 NP_112601.3 8 791
XM_011515437.1 XP_011513739.1 8 958
XM_017012366.1 XP_016867855.1 8 791
GenBank mRNA AF106704.2 - 8 711
AF217508.1 - 7 678
AF282617.1 - 8 741
AK075433.1 - 8 792
AK124472.1 - 12 1319
AK225545.1 - 8 742
AY358549.1 - 8 768
BC016838.2 - 8 839 3888647

View Full Product Details