Gene Transcripts

Gene Symbol
FAM20A
Entrez Gene ID
54757
Gene Name
family with sequence similarity 20 member A
Gene Aliases
AI1G, AIGFS, FP2747
Chromosome Location
Chr. 17: 68534561 - 68600954 on Build GRCh38
UniGene ID
Hs.268874
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243746.1 NP_001230675.1 9 1705
NM_017565.3 NP_060035.2 8 1479
NR_027751.1 8 906
XM_006721959.2 XP_006722022.1 8 851
GenBank mRNA AB545606.1 - 8 1479
AF447872.1 - 1 1116
AK056789.1 - 9 1319
AK298071.1 - 9 1347
AL133105.1 - 6 588
AY358197.1 - 9 1479
BC036222.1 - 8 870
BC136686.1 - 8 1368 9020676
BC136689.1 - 9 1368 9020679

View Full Product Details