Gene Transcripts
- Gene Symbol
- FAM20A
- Entrez Gene ID
- 54757
- Gene Name
- family with sequence similarity 20 member A
- Gene Aliases
- AI1G, AIGFS, FP2747
- Chromosome Location
- Chr. 17: 68534561 - 68600954 on Build GRCh38
- UniGene ID
- Hs.268874
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001243746.1 |
NP_001230675.1
|
11, 12 |
1866 |
|
NM_017565.3 |
NP_060035.2
|
10, 11 |
1640 |
|
NR_027751.1 |
|
10, 11 |
1067 |
|
XM_006721959.2 |
XP_006722022.1
|
10, 11 |
1012 |
|
GenBank mRNA |
AB545606.1 |
- |
10, 11 |
1640 |
|
AF447872.1 |
- |
4, 5 |
1453 |
|
AK056789.1 |
- |
11, 12 |
1480 |
|
AK298071.1 |
- |
11, 12 |
1508 |
|
AL133105.1 |
- |
8, 9 |
749 |
|
AY358197.1 |
- |
11, 12 |
1640 |
|
BC036222.1 |
- |
10, 11 |
1031 |
|
BC136686.1 |
- |
10, 11 |
1529 |
9020676 |
BC136689.1 |
- |
11, 12 |
1529 |
9020679 |
View Full Product Details