Gene Transcripts

Gene Symbol
MBTD1
Entrez Gene ID
54799
Gene Name
mbt domain containing 1
Gene Aliases
SA49P01
Chromosome Location
Chr. 17: 51177419 - 51260511 on Build GRCh38
UniGene ID
Hs.656803
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017643.2 NP_060113.2 13 1590
XM_005257467.4 XP_005257524.1 12 1535
XM_005257468.4 XP_005257525.1 12 1528
XM_011524922.2 XP_011523224.1 12 1537
XM_011524923.2 XP_011523225.1 12 1534
XM_011524924.2 XP_011523226.1 12 1530
XM_011524925.1 XP_011523227.1 13 1651
XM_011524926.2 XP_011523228.1 13 1755
XM_011524928.2 XP_011523230.1 12 1692
XM_011524929.2 XP_011523231.1 13 1606
XM_011524930.1 XP_011523232.1 12 1328
XM_011524931.2 XP_011523233.1 11 1162
XM_011524933.2 XP_011523235.1 14 1712
XM_017024782.1 XP_016880271.1 13 1902
GenBank mRNA AK000062.1 - 9 922
BC034364.1 - 13 1561
BC101736.1 - 8 851 8069242

View Full Product Details