Gene Transcripts

Gene Symbol
HERC6
Entrez Gene ID
55008
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase family member 6
Gene Aliases
Chromosome Location
Chr. 4: 88378565 - 88443097 on Build GRCh38
UniGene ID
Hs.529317
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001165136.1 NP_001158608.1 7 1130
NM_017912.3 NP_060382.3 7 1130
XM_005263083.3 XP_005263140.1 7 1327
XM_011532053.2 XP_011530355.1 7 1329
XM_017008333.1 XP_016863822.1 7 810
GenBank mRNA AF336798.1 - 7 1130
AK295832.1 - 7 1040
AY653201.1 - 7 1130
AY653202.1 - 7 1130
BC042047.1 - 7 1006 5531519
BX647121.1 - 7 806

View Full Product Details