Gene Transcripts
- Gene Symbol
- TMEM255A
- Entrez Gene ID
- 55026
- Gene Name
- transmembrane protein 255A
- Gene Aliases
- FAM70A
- Chromosome Location
- Chr. X: 120250984 - 120311556 on Build GRCh38
- UniGene ID
- Hs.437563
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001104544.1 |
NP_001098014.1
|
2, 3 |
427 |
|
NM_001104545.1 |
NP_001098015.1
|
2, 3 |
427 |
|
NM_017938.3 |
NP_060408.3
|
2, 3 |
427 |
|
XM_017029619.1 |
XP_016885108.1
|
2, 3 |
447 |
|
GenBank mRNA |
AF450484.1 |
- |
2, 3 |
357 |
|
AK056403.1 |
- |
2, 3 |
430 |
|
AK289684.1 |
- |
2, 3 |
318 |
|
BC047054.1 |
- |
2, 3 |
324 |
5313196 |
DA758358.1 |
- |
2, 3 |
428 |
|
DB165640.1 |
- |
2, 3 |
324 |
|
View Full Product Details