Gene Transcripts
- Gene Symbol
- CEP55
- Entrez Gene ID
- 55165
- Gene Name
- centrosomal protein 55
- Gene Aliases
- C10orf3, CT111, URCC6
- Chromosome Location
- Chr. 10: 93496612 - 93529094 on Build GRCh38
- UniGene ID
- Hs.14559
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127182.1 |
NP_001120654.1
|
7, 8 |
1248 |
|
NM_018131.4 |
NP_060601.3
|
7, 8 |
1370 |
|
XM_011539918.1 |
XP_011538220.1
|
7, 8 |
1384 |
|
XM_011539919.1 |
XP_011538221.1
|
7, 8 |
1252 |
|
XM_011539920.2 |
XP_011538222.1
|
7, 8 |
1078 |
|
XM_017016372.1 |
XP_016871861.1
|
7, 8 |
1224 |
|
XM_017016373.1 |
XP_016871862.1
|
7, 8 |
1246 |
|
GenBank mRNA |
AB091343.1 |
- |
7, 8 |
1359 |
|
AK001402.1 |
- |
6, 7 |
970 |
|
AK223495.1 |
- |
7, 8 |
1351 |
|
AK315536.1 |
- |
7, 8 |
1367 |
|
AY788918.1 |
- |
7, 8 |
1370 |
|
BC008947.2 |
- |
7, 8 |
1350 |
3028566 |
BX640718.1 |
- |
8, 9 |
1290 |
|
View Full Product Details