Gene Transcripts

Gene Symbol
USP40
Entrez Gene ID
55230
Gene Name
ubiquitin specific peptidase 40
Gene Aliases
Chromosome Location
Chr. 2: 233475519 - 233566789 on Build GRCh38
UniGene ID
Hs.96513
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018218.2 NP_060688.1 14, 15 1922
XM_006712612.3 XP_006712675.1 14, 15 1940
XM_011511396.2 XP_011509698.1 14, 15 1940
XM_011511397.2 XP_011509699.1 15, 16 2014
XM_011511398.2 XP_011509700.1 14, 15 1940
XM_011511399.2 XP_011509701.1 13, 14 1741
XM_011511400.2 XP_011509702.1 13, 14 1725
XM_011511401.2 XP_011509703.1 14, 15 1940
XM_017004426.1 XP_016859915.1 15, 16 2014
XM_017004427.1 XP_016859916.1 8, 9 942
XM_017004428.1 XP_016859917.1 14, 15 1982
GenBank mRNA AJ583821.2 - 14, 15 1922
AK001647.1 - 14, 15 1884
AK297821.1 - 8, 9 1010

View Full Product Details