Gene Transcripts
- Gene Symbol
- USP40
- Entrez Gene ID
- 55230
- Gene Name
- ubiquitin specific peptidase 40
- Gene Aliases
- Chromosome Location
- Chr. 2: 233475519 - 233566789 on Build GRCh38
- UniGene ID
- Hs.96513
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018218.2 |
NP_060688.1
|
26, 27 |
3160 |
|
XM_006712612.3 |
XP_006712675.1
|
26, 27 |
3178 |
|
XM_011511396.2 |
XP_011509698.1
|
26, 27 |
3190 |
|
XM_011511397.2 |
XP_011509699.1
|
27, 28 |
3264 |
|
XM_011511398.2 |
XP_011509700.1
|
26, 27 |
3190 |
|
XM_011511399.2 |
XP_011509701.1
|
25, 26 |
2991 |
|
XM_011511400.2 |
XP_011509702.1
|
25, 26 |
2975 |
|
XM_017004426.1 |
XP_016859915.1
|
27, 28 |
3252 |
|
XM_017004427.1 |
XP_016859916.1
|
20, 21 |
2192 |
|
XM_017004428.1 |
XP_016859917.1
|
26, 27 |
3232 |
|
GenBank mRNA |
AJ583821.2 |
- |
26, 27 |
3160 |
|
AK001647.1 |
- |
26, 27 |
3122 |
|
AK297821.1 |
- |
20, 21 |
2260 |
|
AL833371.1 |
- |
1, 2 |
272 |
|
View Full Product Details