Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018518.4 | NP_060988.3 | 9 | 1266 | |
NM_182751.2 | NP_877428.1 | 9 | 1269 | ||
XM_011519538.2 | XP_011517840.1 | 9 | 1239 | ||
GenBank mRNA | AB042719.1 | - | 9 | 1233 | |
AK055695.1 | - | 9 | 1242 | ||
AK292701.1 | - | 9 | 1244 | ||
AL136840.1 | - | 8 | 1232 | ||
BC004876.2 | - | 2 | 184 | ||
BC009108.1 | - | 4 | 484 | ||
BC101727.1 | - | 9 | 1238 | 8069233 | |
BC143490.1 | - | 9 | 1231 | 9051998 |