Gene Transcripts

Gene Symbol
POMGNT1
Entrez Gene ID
55624
Gene Name
protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-)
Gene Aliases
GNTI.2, GnT I.2, LGMD2O, MEB, MGAT1.2, gnT-I.2
Chromosome Location
Chr. 1: 46188681 - 46220305 on Build GRCh38
UniGene ID
Hs.525134
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243766.1 NP_001230695.1 20 2334
NM_001290129.1 NP_001277058.1 19 1755
NM_001290130.1 NP_001277059.1 19 1721
NM_017739.3 NP_060209.3 20 1847
XM_005271010.1 XP_005271067.1 20 1842
XM_006710755.1 XP_006710818.1 20 1807
XM_006710756.1 XP_006710819.1 20 1842
XM_011541760.2 XP_011540062.1 19 1699
XM_017001690.1 XP_016857179.1 20 1807
GenBank mRNA AB057356.1 - 20 1777
AF250859.2 - 17 1436
AK000284.1 - 20 1824
AK022727.1 - 15 1193
AK056186.1 - 20 2334
AK225033.1 - 20 1825
AK301923.1 - 19 1721
AK302379.1 - 19 1731
AK316399.1 - 19 1728
AW818474.1 - 2 202
AY358592.1 - 20 1812
BC001471.2 - 20 1841 3138971
CR749700.1 - 17 2592

View Full Product Details