Gene Transcripts

Gene Symbol
MFN1
Entrez Gene ID
55669
Gene Name
mitofusin 1
Gene Aliases
hfzo1, hfzo2
Chromosome Location
Chr. 3: 179347692 - 179394944 on Build GRCh38
UniGene ID
Hs.478383
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033540.2 NP_284941.2 9 1061
XM_005247596.3 XP_005247653.2 9 1019
XM_011512963.2 XP_011511265.1 6 647
GenBank mRNA AB043588.1 - 8 1026
AB043589.1 - 9 1026
AF329637.1 - 8 935
AK000700.1 - 9 1018
AK314306.1 - 9 1018
BC020661.1 - 9 1027
BC040557.1 - 9 1061 5270347
CR627379.1 - 6 711
U95822.1 - 5 524

View Full Product Details