Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018448.4 | NP_060918.2 | 1, 2 | 507 | |
GenBank mRNA | AB020636.2 | - | 1, 2 | 409 | |
AK096010.1 | - | 1, 2 | 413 | ||
AK292456.1 | - | 3, 4 | 346 | ||
AK314358.1 | - | 1, 2 | 395 | ||
AL136810.1 | - | 1, 2 | 419 | ||
BC050341.1 | - | 1, 2 | 203 | 5265409 | |
DB064887.1 | - | 1, 2 | 509 | ||
DB098973.1 | - | 1, 2 | 484 |