Gene Transcripts

Gene Symbol
HMCES
Entrez Gene ID
56941
Gene Name
5-hydroxymethylcytosine (hmC) binding, ES cell-specific
Gene Aliases
C3orf37, DC12, SRAPD1
Chromosome Location
Chr. 3: 129278806 - 129305293 on Build GRCh38
UniGene ID
Hs.458320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006109.1 NP_001006109.1 6 1022
NM_020187.2 NP_064572.2 6 849
XM_005247636.3 XP_005247693.1 6 1034
XM_005247637.3 XP_005247694.1 5 758
XM_017006877.1 XP_016862366.1 6 866
GenBank mRNA AF201934.1 - 6 862
AK025883.1 - 5 694
AK225264.1 - 5 718
AK297501.1 - 5 721
AK309891.1 - 6 846
AY007144.1 - 2 300
BC009993.2 - 6 929 4130099
BC010125.2 - 6 812 3945258
BC050686.1 - 6 1022 6155077
BC088363.1 - 6 846 30398247

View Full Product Details