Gene Transcripts
- Gene Symbol
- RAB40C
- Entrez Gene ID
- 57799
- Gene Name
- RAB40C, member RAS oncogene family
- Gene Aliases
- RARL, RASL8C
- Chromosome Location
- Chr. 16: 589357 - 629273 on Build GRCh38
- UniGene ID
- Hs.459630
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001172663.1 |
NP_001166134.1
|
2, 3 |
375 |
|
NM_001172664.1 |
NP_001166135.1
|
2, 3 |
267 |
|
NM_001172665.1 |
NP_001166136.1
|
2, 3 |
227 |
|
NM_001172666.1 |
NP_001166137.1
|
2, 3 |
227 |
|
NM_021168.4 |
NP_066991.3
|
1, 2 |
272 |
|
GenBank mRNA |
AY823398.1 |
- |
1, 2 |
197 |
|
BC028696.1 |
- |
1, 2 |
274 |
4826800 |
BG696681.1 |
- |
2, 3 |
376 |
|
BG715678.1 |
- |
2, 3 |
270 |
|
BM545442.1 |
- |
1, 2 |
346 |
|
BQ685517.1 |
- |
2, 3 |
206 |
|
CX783728.1 |
- |
2, 3 |
360 |
|
View Full Product Details