Gene Transcripts
- Gene Symbol
- LHPP
- Entrez Gene ID
- 64077
- Gene Name
- phospholysine phosphohistidine inorganic pyrophosphate phosphatase
- Gene Aliases
- HDHD2B
- Chromosome Location
- Chr. 10: 124461772 - 124614141 on Build GRCh38
- UniGene ID
- Hs.527748
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001167880.1 |
NP_001161352.1
|
2, 3 |
407 |
|
NM_001318331.1 |
NP_001305260.1
|
2, 3 |
407 |
|
NM_001318332.1 |
NP_001305261.1
|
2, 3 |
407 |
|
NM_022126.3 |
NP_071409.3
|
2, 3 |
407 |
|
XM_005270026.3 |
XP_005270083.1
|
2, 3 |
356 |
|
XM_006717943.1 |
XP_006718006.1
|
2, 3 |
356 |
|
XM_011540058.2 |
XP_011538360.1
|
2, 3 |
356 |
|
XM_017016509.1 |
XP_016871998.1
|
2, 3 |
356 |
|
XM_017016510.1 |
XP_016871999.1
|
2, 3 |
356 |
|
XM_017016511.1 |
XP_016872000.1
|
2, 3 |
356 |
|
GenBank mRNA |
AK055532.1 |
- |
2, 3 |
336 |
|
AW631352.1 |
- |
2, 3 |
318 |
|
BC110344.1 |
- |
2, 3 |
361 |
5762731 |
BC113629.1 |
- |
2, 3 |
407 |
8322681 |
BC113631.1 |
- |
2, 3 |
407 |
8322683 |
BM541880.1 |
- |
2, 3 |
345 |
|
HY154569.1 |
- |
2, 3 |
371 |
|
View Full Product Details