Gene Transcripts

Gene Symbol
CHST8
Entrez Gene ID
64377
Gene Name
carbohydrate sulfotransferase 8
Gene Aliases
GALNAC4ST1, GalNAc4ST, PSS3
Chromosome Location
Chr. 19: 33621955 - 33773509 on Build GRCh38
UniGene ID
Hs.165724
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127895.1 NP_001121367.1 5 1339
NM_001127896.1 NP_001121368.1 4 1250
NM_022467.3 NP_071912.2 4 1592
XM_011527222.1 XP_011525524.1 4 1573
XM_011527224.1 XP_011525526.1 5 1707
XM_017027143.1 XP_016882632.1 4 1243
GenBank mRNA AB047801.1 - 4 1106
AF300612.1 - 4 1053
AF305781.1 - 4 1149
BC011380.1 - 4 1202 3874578
BC014250.2 - 4 1246 4635405
BC018723.1 - 4 1592 4137487

View Full Product Details