Gene Transcripts

Gene Symbol
ALDH8A1
Entrez Gene ID
64577
Gene Name
aldehyde dehydrogenase 8 family member A1
Gene Aliases
ALDH12, DJ352A20.2
Chromosome Location
Chr. 6: 134917390 - 134950122 on Build GRCh38
UniGene ID
Hs.486520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022568.3 NP_072090.1 4 642
NM_170771.2 NP_739577.1 4 642
GenBank mRNA AF303134.1 - 4 633
AK074266.1 - 4 635
AK222848.1 - 4 635
AK290784.1 - 4 589
BC113862.1 - 4 612 40033493
BC114473.1 - 5 650 40033488

View Full Product Details