Gene Transcripts

Gene Symbol
GORASP1
Entrez Gene ID
64689
Gene Name
golgi reassembly stacking protein 1
Gene Aliases
GOLPH5, GRASP65, P65
Chromosome Location
Chr. 3: 39096599 - 39108363 on Build GRCh38
UniGene ID
Hs.721946
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031899.3 NP_114105.1 3 993
NR_103866.1 3 530
NR_103867.1 3 530
XM_006713301.3 XP_006713364.1 3 1093
XM_011534020.2 XP_011532322.1 3 1093
XM_011534021.2 XP_011532323.1 3 1093
XM_017007050.1 XP_016862539.1 3 269
XM_017007051.1 XP_016862540.1 3 708
XM_017007052.1 XP_016862541.1 3 255
GenBank mRNA AJ409349.1 - 3 269
AK001574.1 - 3 268
AK091168.1 - 3 457
AK124755.1 - 3 993
AK300271.1 - 3 209
BC008928.2 - 3 251
BC075854.1 - 3 279 5503263
BC103826.1 - 3 211 40001584
BC103827.1 - 3 211 40001588
DB031221.1 - 3 268

View Full Product Details