Gene Transcripts

Gene Symbol
FKRP
Entrez Gene ID
79147
Gene Name
fukutin related protein
Gene Aliases
LGMD2I, MDC1C, MDDGA5, MDDGB5, MDDGC5
Chromosome Location
Chr. 19: 46746015 - 46758575 on Build GRCh38
UniGene ID
Hs.515493
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039885.2 NP_001034974.1 4 1574
NM_024301.4 NP_077277.1 4 1522
XM_005259247.2 XP_005259304.1 5 1635
XM_005259248.2 XP_005259305.1 5 1625
XM_005259249.4 XP_005259306.1 5 1567
XM_011527306.1 XP_011525608.1 3 1371
XM_011527307.1 XP_011525609.1 4 1420
XM_017027297.1 XP_016882786.1 4 1741
GenBank mRNA AJ314847.1 - 4 1510
AK022638.1 - 4 1510
AK095497.1 - 4 1561
AK291282.1 - 4 1510
BC002612.2 - 2 1318 3160297

View Full Product Details