Gene Transcripts

Gene Symbol
TMEM43
Entrez Gene ID
79188
Gene Name
transmembrane protein 43
Gene Aliases
ARVC5, ARVD5, EDMD7, LUMA
Chromosome Location
Chr. 3: 14124940 - 14143680 on Build GRCh38
UniGene ID
Hs.517817
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024334.2 NP_077310.1 11 1216
XM_017007176.1 XP_016862665.1 12 1338
GenBank mRNA AK027466.1 - 5 446
AK027757.1 - 11 1142
AK027827.1 - 6 499
AK027877.1 - 11 1080
AK074073.1 - 9 3647
AK075010.1 - 11 1067
AK225589.1 - 11 1139
AL136916.1 - 11 1110
AY358625.1 - 11 979
BC003125.1 - 11 1111 3503636
BC008054.2 - 9 706
BC011719.2 - 11 1111 4079766
KU178769.1 - 10 962
KU178770.1 - 9 1536

View Full Product Details