Gene Transcripts
- Gene Symbol
- GLB1L
- Entrez Gene ID
- 79411
- Gene Name
- galactosidase beta 1 like
- Gene Aliases
- Chromosome Location
- Chr. 2: 219236598 - 219245429 on Build GRCh38
- UniGene ID
- Hs.181173
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286423.1 |
NP_001273352.1
|
12, 13 |
1406 |
|
NM_001286427.1 |
NP_001273356.1
|
9, 10 |
923 |
|
NM_024506.4 |
NP_078782.3
|
12, 13 |
1394 |
|
XM_011511813.2 |
XP_011510115.1
|
10, 11 |
983 |
|
XM_017004895.1 |
XP_016860384.1
|
10, 11 |
1090 |
|
GenBank mRNA |
AK055098.1 |
- |
7, 8 |
581 |
|
AK074393.1 |
- |
6, 7 |
574 |
|
AK299404.1 |
- |
11, 12 |
1155 |
|
AK300215.1 |
- |
9, 10 |
906 |
|
AY358639.1 |
- |
12, 13 |
1374 |
|
View Full Product Details