Gene Transcripts

Gene Symbol
HAUS3
Entrez Gene ID
79441
Gene Name
HAUS augmin like complex subunit 3
Gene Aliases
C4orf15, IT1, dgt3
Chromosome Location
Chr. 4: 2231836 - 2242164 on Build GRCh38
UniGene ID
Hs.665869
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303143.1 NP_001290072.1 3 1098
NM_024511.6 NP_078787.2 2 827
GenBank mRNA AF040964.1 - 1 571
AK056134.1 - 2 1315
AK293948.1 - 2 1315
BC003648.2 - 3 1099 3534250
BC025356.1 - 1 339 4906369
CR749640.1 - 2 796

View Full Product Details