Gene Transcripts

Gene Symbol
PANK3
Entrez Gene ID
79646
Gene Name
pantothenate kinase 3
Gene Aliases
Chromosome Location
Chr. 5: 168555623 - 168579609 on Build GRCh38
UniGene ID
Hs.388400
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024594.3 NP_078870.1 3 928
GenBank mRNA AK022961.1 - 3 673
AK223285.1 - 3 657
BC013705.1 - 3 902 4343270

View Full Product Details