Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001114133.2 | NP_001107605.1 | 4 | 2715 | |
NM_024875.4 | NP_079151.2 | 2 | 1892 | ||
XM_005270158.2 | XP_005270215.1 | 2 | 2214 | ||
XM_005270159.3 | XP_005270216.1 | 2 | 1848 | ||
GenBank mRNA | AB188489.1 | - | 4 | 2684 | |
AK022983.1 | - | 2 | 1882 | ||
AK295873.1 | - | 4 | 2604 | ||
BC142635.1 | - | 2 | 1874 | 40147769 |