Gene Transcripts

Gene Symbol
WLS
Entrez Gene ID
79971
Gene Name
wntless Wnt ligand secretion mediator
Gene Aliases
C1orf139, EVI, GPR177, MRP, mig-14
Chromosome Location
Chr. 1: 68098459 - 68233000 on Build GRCh38
UniGene ID
Hs.647659
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002292.3 NP_001002292.3 3 678
NM_001193334.1 NP_001180263.1 2 411
NM_024911.6 NP_079187.3 3 739
XM_011542191.2 XP_011540493.1 3 662
XM_011542192.2 XP_011540494.1 3 672
XM_017002390.1 XP_016857879.1 3 506
GenBank mRNA AK074583.1 - 3 694
AK074984.1 - 3 661
AK301613.1 - 2 388
AK309779.1 - 4 618
BX648748.1 - 2 409
DQ323735.1 - 3 708

View Full Product Details