Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253875.1 | NP_001240804.1 | 15 | 1257 | |
NM_001253876.1 | NP_001240805.1 | 10 | 1394 | ||
NM_025076.4 | NP_079352.2 | 15 | 1242 | ||
NR_045607.1 | 14 | 1193 | |||
XM_006712775.3 | XP_006712838.1 | 14 | 1115 | ||
XM_011511902.2 | XP_011510204.2 | 15 | 1236 | ||
XM_011511903.2 | XP_011510205.2 | 15 | 1219 | ||
XM_011511904.2 | XP_011510206.1 | 12 | 1564 | ||
XM_017005008.1 | XP_016860497.1 | 13 | 1057 | ||
XM_017005009.1 | XP_016860498.1 | 13 | 1042 | ||
XM_017005010.1 | XP_016860499.1 | 14 | 1100 | ||
XM_017005011.1 | XP_016860500.1 | 15 | 1344 | ||
XM_017005012.1 | XP_016860501.1 | 15 | 1279 | ||
XM_017005013.1 | XP_016860502.1 | 13 | 1802 | ||
XM_017005014.1 | XP_016860503.1 | 14 | 1020 | ||
GenBank mRNA | AK027244.1 | - | 10 | 1394 | |
AK056022.1 | - | 4 | 2071 | ||
AK075120.1 | - | 15 | 1204 | ||
AK075170.1 | - | 15 | 1219 | ||
AK122696.1 | - | 14 | 1158 | ||
AK225592.1 | - | 10 | 1394 | ||
AK290668.1 | - | 15 | 1217 | ||
AL834439.1 | - | 2 | 1006 | ||
AY147934.1 | - | 15 | 1209 | ||
AY358541.1 | - | 15 | 1193 | ||
BC009819.2 | - | 15 | 1183 | 3843312 | |
BX366726.2 | - | 1 | 817 |