Gene Transcripts

Gene Symbol
SP6
Entrez Gene ID
80320
Gene Name
Sp6 transcription factor
Gene Aliases
EPFN, EPIPROFIN, KLF14
Chromosome Location
Chr. 17: 47844908 - 47876312 on Build GRCh38
UniGene ID
Hs.253603
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258248.1 NP_001245177.1 2 1046
NM_199262.2 NP_954871.1 2 1035
XM_006722115.3 XP_006722178.1 2 998
GenBank mRNA AK127707.1 - 2 1089
AK127850.1 - 2 1035
BC103951.1 - 2 934 40011121
BC103952.1 - 2 934 40011125
BC103953.1 - 2 934 40011129
BC103954.1 - 2 934 40011132

View Full Product Details