Gene Transcripts

Gene Symbol
RTBDN
Entrez Gene ID
83546
Gene Name
retbindin
Gene Aliases
Chromosome Location
Chr. 19: 12825477 - 12835428 on Build GRCh38
UniGene ID
Hs.21162
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080997.2 NP_001074466.1 3 381
NM_001270440.1 NP_001257369.1 4 654
NM_001270441.1 NP_001257370.1 3 334
NM_001270442.1 NP_001257371.1 3 322
NM_001270443.1 NP_001257372.1 3 310
NM_001270444.1 NP_001257373.1 3 298
NM_001270445.1 NP_001257374.1 3 292
NM_031429.2 NP_113617.1 4 654
XM_005260089.1 XP_005260146.1 4 729
XM_005260090.3 XP_005260147.1 3 592
XM_011528343.1 XP_011526645.1 3 395
XM_011528344.2 XP_011526646.1 3 445
XM_011528345.1 XP_011526647.1 3 390
XM_017027343.1 XP_016882832.1 4 684
XM_017027344.1 XP_016882833.1 4 636
GenBank mRNA AB593122.1 - 3 334
AK093672.1 - 4 654
AY028917.1 - 3 358
BC005063.1 - 4 642 3344818
BG471112.1 - 3 295
BM688006.1 - 3 286
BM688127.1 - 3 287
BM708463.1 - 3 381
BQ639040.1 - 4 256
BU689643.1 - 4 704
EL946853.1 - 3 314

View Full Product Details