Gene Transcripts

Gene Symbol
PARP9
Entrez Gene ID
83666
Gene Name
poly(ADP-ribose) polymerase family member 9
Gene Aliases
ARTD9, BAL, BAL1, MGC:7868
Chromosome Location
Chr. 3: 122527910 - 122564676 on Build GRCh38
UniGene ID
Hs.518200
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146102.1 NP_001139574.1 4 998
NM_001146103.1 NP_001139575.1 4 1075
NM_001146104.1 NP_001139576.1 4 1074
NM_001146105.1 NP_001139577.1 4 893
NM_001146106.1 NP_001139578.1 4 1074
NM_031458.2 NP_113646.2 4 1180
XM_005247820.2 XP_005247877.1 4 1104
XM_011513218.1 XP_011511520.1 4 970
XM_017007304.1 XP_016862793.1 4 988
XM_017007305.1 XP_016862794.1 4 883
XM_017007306.1 XP_016862795.1 4 964
GenBank mRNA AF307338.1 - 4 1081
AF307339.1 - 4 976
AK292959.1 - 4 853
AK299107.1 - 4 967
AK313494.1 - 4 967
BC017463.1 - 4 1042
BC039580.1 - 3 851 5416494

View Full Product Details