Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300948.1 | NP_001287877.1 | 2 | 543 | |
NM_031490.3 | NP_113678.2 | 2 | 543 | ||
XM_017023755.1 | XP_016879244.1 | 2 | 410 | ||
XM_017023756.1 | XP_016879245.1 | 2 | 408 | ||
GenBank mRNA | AJ548761.1 | - | 2 | 410 | |
AK027544.1 | - | 2 | 374 | ||
AK074775.1 | - | 2 | 408 | ||
AK127125.1 | - | 3 | 306 | ||
AK294779.1 | - | 2 | 355 | ||
BC093910.1 | - | 2 | 377 | 7939755 | |
BC093912.1 | - | 2 | 377 | 7939757 | |
BC143246.1 | - | 2 | 377 | 9051747 | |
DC316485.1 | - | 2 | 441 |